Learn More
Biomatik Corporation Recombinant Human Desmocollin 1 (DSC1)
Supplier:  Biomatik Corporation RPU41175
Encompass_Preferred
Recombinant Human Desmocollin 1 (DSC1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Desmocollin 1 (DSC1). Target Synonyms: CDHF1; DG2/DG3; Cadherin family member 1; Desmosomal glycoprotein 2/3. Accession Number: Q08554. Gene ID: 1823. Expression Region: Ala486~Asn686. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9925