Learn More
Biomatik Corporation Recombinant Mouse Ferritin, Heavy Polypeptide (FTH)
Supplier:  Biomatik Corporation RPU41388
Encompass_Preferred
Recombinant Mouse Ferritin, Heavy Polypeptide (FTH) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Ferritin, Heavy Polypeptide (FTH). Target Synonyms: FTH1; FTHL6; PIG15; PLIF; Ferritin Heavy Chain; Apoferritin; Placenta Immunoregulatory Factor; Proliferation-Inducing Protein 15; Cell proliferation-inducing gene 15 protein. Accession Number: P09528. Gene ID: 14319. Expression Region: Met1~Ser182. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9941