Learn More
Biomatik Corporation Recombinant Mouse Fibroblast Growth Factor 10 (FGF10)
Supplier:  Biomatik Corporation RPU41435
Encompass_Preferred
Recombinant Mouse Fibroblast Growth Factor 10 (FGF10) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Fibroblast Growth Factor 10 (FGF10). Target Synonyms: Keratinocyte growth factor 2. Accession Number: O35565. Gene ID: 14165. Expression Region: Gln37~Thr209. Tag Info: N-terminal His and GST Tag. Theoretical MW: 50kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9943