Learn More
Biomatik Corporation Recombinant Human Fibroblast Growth Factor Receptor 3 (FGFR3)
Supplier:  Biomatik Corporation RPU41453
Encompass_Preferred
Recombinant Human Fibroblast Growth Factor Receptor 3 (FGFR3) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Fibroblast Growth Factor Receptor 3 (FGFR3). Target Synonyms: CD333; ACH; CEK2; JTK4; Achondroplasia; Thanatophoric Dwarfism. Accession Number: P22607. Gene ID: 2261. Expression Region: Val166~Gly375. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9947