Learn More
Biomatik Corporation Recombinant Human Galectin 1 (GAL1)
Supplier:  Biomatik Corporation RPU41557
Encompass_Preferred
Recombinant Human Galectin 1 (GAL1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Galectin 1 (GAL1). Target Synonyms: LGALS1; GBP; HLBP14; Galaptin; Lectin,Galactoside-Binding Soluble 1; 14 kDa laminin-binding protein; Lactose-binding lectin 1; Putative MAPK-activating protein PM12; S-Lac lectin. Accession Number: P09382. Gene ID: 3956. Expression Region: Met1~Asp135. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9952