Learn More
Biomatik Corporation Recombinant Mouse Galectin 9 (GAL9)
Supplier:  Biomatik Corporation RPU41572
Encompass_Preferred
Recombinant Mouse Galectin 9 (GAL9) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Galectin 9 (GAL9). Target Synonyms: LGALS9; Ecalectin; Lectin,Galactoside-Binding Soluble 9; Tumor antigen HOM-HD-21. Accession Number: O08573. Gene ID: 16859. Expression Region: Phe17~Gln147. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9954