Learn More
Biomatik Corporation Recombinant Human Granulin (GRN)
Supplier:  Biomatik Corporation RPU41729
Encompass_Preferred
Recombinant Human Granulin (GRN) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Granulin (GRN). Target Synonyms: GEP; GP88; PCDGF; PEPI; PGRN; Proepithelin; Acrogranin; Glycoprotein of 88 Kda; Paragranulin. Accession Number: P28799. Gene ID: 2896. Expression Region: Lys361~Arg585. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9963