Learn More
Biomatik Corporation Recombinant Human Growth Differentiation Factor 15 (GDF15)
Supplier:  Biomatik Corporation RPU41760
Encompass_Preferred
Recombinant Human Growth Differentiation Factor 15 (GDF15) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Growth Differentiation Factor 15 (GDF15). Target Synonyms: PDF; MIC1; NAG-1; PLAB; PTGFB; Macrophage inhibitory cytokine 1; NSAID-activated gene 1 protein; Placental bone morphogenetic protein; Prostate differentiation factor. Accession Number: Q99988. Gene ID: 9518. Expression Region: Ala195~Ile308. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9967