Learn More
Biomatik Corporation Recombinant Human Heme Oxygenase 2, Decycling (HO2)
Supplier:  Biomatik Corporation RPU41851
Encompass_Preferred
Recombinant Human Heme Oxygenase 2, Decycling (HO2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Heme Oxygenase 2, Decycling (HO2). Target Synonyms: HMOX2. Accession Number: P30519. Gene ID: 3163. Expression Region: Thr71~Leu306. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9972