Learn More
Biomatik Corporation Recombinant Human Inhibin Beta B (INHbB)
Supplier:  Biomatik Corporation RPU42000
Encompass_Preferred
Recombinant Human Inhibin Beta B (INHbB) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Inhibin Beta B (INHbB). Target Synonyms: INH-BB; Activin AB Beta Polypeptide. Accession Number: P09529. Gene ID: 3625. Expression Region: Gly293~Ala407. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9980