Learn More
Biomatik Corporation Recombinant Human Neutrophil Elastase (NE)
Supplier:  Biomatik Corporation RPU41245
Encompass_Preferred
Recombinant Human Neutrophil Elastase (NE) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Neutrophil Elastase (NE). Target Synonyms: ELANE; HLE; HNE; ELA2; PMN-E; Neutrophil Elastase; Medullasin; Polymorphonuclear Leukocyte Elastase; Bone Marrow Serine Protease; Human Leukocyte Elastase; PMN Elastase. Accession Number: P08246. Gene ID: 1991. Expression Region: Ile30~Gln247. Tag Info: N-terminal His and GST Tag. Theoretical MW: 53/28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9930