Learn More
Biomatik Corporation Recombinant Mouse Interleukin 18 Binding Protein (IL18BP)
Supplier:  Biomatik Corporation RPU42258
Encompass_Preferred
Recombinant Mouse Interleukin 18 Binding Protein (IL18BP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Interleukin 18 Binding Protein (IL18BP). Target Synonyms: IL18BPa; MC51L-53L-54L Homolog Gene Product; Tadekinig-alfa. Accession Number: Q9Z0M9. Gene ID: 16068. Expression Region: Thr29~Ala193. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9990