Learn More
Biomatik Corporation Recombinant Human Cytokeratin 7 (CK7)
Supplier:  Biomatik Corporation RPU42401
Encompass_Preferred
Recombinant Human Cytokeratin 7 (CK7) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cytokeratin 7 (CK7). Target Synonyms: KRT7; K2C7; K7; SCL; Keratin 7; Sarcolectin; Keratin,55K Type II Cytoskeletal; Keratin, type II cytoskeletal 7. Accession Number: P08729. Gene ID: 3855. Expression Region: Glu91~Lys394. Tag Info: N-terminal His Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9999