Learn More
Biomatik Corporation Recombinant Human Leucine Aminopeptidase (LAP)
Supplier:  Biomatik Corporation RPU42510
Encompass_Preferred
Recombinant Human Leucine Aminopeptidase (LAP) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Leucine Aminopeptidase (LAP). Target Synonyms: LAPEP, LAP3; PEPS; Peptidase S; Leucyl Aminopeptidase; Cytosol aminopeptidase; Proline aminopeptidase; Prolyl aminopeptidase. Accession Number: P28838. Gene ID: 51056. Expression Region: Ala320~Ala519. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0004