Learn More
Biomatik Corporation Recombinant Human Epidermal Growth Factor Receptor (EGFR)
Supplier:  Biomatik Corporation RPU41298
Encompass_Preferred
Recombinant Human Epidermal Growth Factor Receptor (EGFR) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Epidermal Growth Factor Receptor (EGFR). Target Synonyms: ErbB-1; ErbB1; ER1; ERB-B1; MENA; HER1; Erythroblastic Leukemia Viral(v-Erb-B)oncogene Homolog,Avian; Proto-oncogene c-ErbB-1; Receptor tyrosine-protein kinase erbB-1. Accession Number: P00533. Gene ID: 1956. Expression Region: Lys713~Asp984. Tag Info: N-terminal His Tag. Theoretical MW: 37kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9936