Learn More
Biomatik Corporation Recombinant Mouse Mucosal Addressin Cell Adhesion Molecule 1 (MAdCAM1)
Supplier:  Biomatik Corporation RPU42841
Encompass_Preferred
Recombinant Mouse Mucosal Addressin Cell Adhesion Molecule 1 (MAdCAM1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Mucosal Addressin Cell Adhesion Molecule 1 (MAdCAM1). Target Synonyms: Addressin. Accession Number: Q61826. Expression Region: Pro29~Cys297. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0020