Learn More
Biomatik Corporation Recombinant Human Neurofilament, Light Polypeptide (NEFL)
Supplier:  Biomatik Corporation RPU43009
Encompass_Preferred
Recombinant Human Neurofilament, Light Polypeptide (NEFL) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Neurofilament, Light Polypeptide (NEFL). Target Synonyms: CMT1F; CMT2E; NF-L; NF68; NFL; 68 kDa neurofilament protein; Neurofilament triplet L protein. Accession Number: P07196. Gene ID: 4747. Expression Region: Lys281~Glu396. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0024