Learn More
Biomatik Corporation Recombinant Mouse Orosomucoid 2 (ORM2)
Supplier:  Biomatik Corporation RPU43150
Encompass_Preferred
Recombinant Mouse Orosomucoid 2 (ORM2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Orosomucoid 2 (ORM2). Target Synonyms: AGP-B; AGP2; Alpha-1-Acid Glycoprotein Type 2. Accession Number: P07361. Gene ID: 18406. Expression Region: Gln19~Ala207. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0031