Learn More
Biomatik Corporation Recombinant Human Phenylalanine Hydroxylase (PAH)
Supplier:  Biomatik Corporation RPU43261
Encompass_Preferred
Recombinant Human Phenylalanine Hydroxylase (PAH) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Phenylalanine Hydroxylase (PAH). Target Synonyms: PKU; PKU1; Phe-4-monooxygenase. Accession Number: P00439. Gene ID: 5053. Expression Region: Met1~Gln226. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0038