Learn More
Biomatik Corporation Recombinant Mouse Phospholipase A2 Group VII (LpPLA2)
Supplier:  Biomatik Corporation RPU43292
Encompass_Preferred
Recombinant Mouse Phospholipase A2 Group VII (LpPLA2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Phospholipase A2 Group VII (LpPLA2). Target Synonyms: PLA2G7; PAF-AH; PAFAH; Lp-PLA2; LDL-PLA2; Platelet Activating Factor Acetylhydrolase,Plasma; Phospholipase A2,Group VII; LDL-associated phospholipase A2; Phospholipase A2, Lipoprotein Associated. Accession Number: Q60963. Gene ID: 27226. Expression Region: Phe22~Asn440. Tag Info: N-terminal His Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0041