Learn More
Biomatik Corporation Recombinant Human Endoplasmic Reticulum Resident Protein 57 (ERp57)
Supplier:  Biomatik Corporation RPU43480
Encompass_Preferred
Recombinant Human Endoplasmic Reticulum Resident Protein 57 (ERp57) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Endoplasmic Reticulum Resident Protein 57 (ERp57). Target Synonyms: P58; ERp61; PDIA3; ERp60; GRP57; PI-PLC; GRP58; Glucose Regulated Protein,58kDa; Protein Disulfide Isomerase-Associated 3; Endoplasmic reticulum resident protein 57. Accession Number: P30101. Gene ID: 2923. Expression Region: Asp162~Tyr416. Tag Info: N-terminal His Tag. Theoretical MW: 33/24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0049