Learn More
Biomatik Corporation Recombinant Human Cluster of Differentiation 90 (CD90)
Supplier:  Biomatik Corporation RPU44000
Encompass_Preferred
Recombinant Human Cluster of Differentiation 90 (CD90) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cluster of Differentiation 90 (CD90). Target Synonyms: Thy1; CDw90; THY mocyte differentiation antigen 1. Accession Number: P04216. Gene ID: 7070. Expression Region: Gln20~Ile133. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0066