Learn More
Biomatik Corporation Native Simian Albumin (ALB)
Supplier:  Biomatik Corporation RPU50320
Encompass_Preferred
Native Simian Albumin (ALB) is a purified Natural Protein. Purity: >90% by SDS-PAGE. Host: Simian. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Simian. Target Name: Albumin (ALB). Target Synonyms: Serum albumin. Accession Number: Q28522. Gene ID: 704892. Theoretical MW: 69kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0093