Learn More
Biomatik Corporation Recombinant Human Riboflavin Kinase (RFK)
Supplier:  Biomatik Corporation RPU50402
Encompass_Preferred
Recombinant Human Riboflavin Kinase (RFK) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Riboflavin Kinase (RFK). Target Synonyms: RIFK; Flavokinase; ATP:riboflavin 5'-phosphotransferase. Accession Number: Q969G6. Gene ID: 55312. Expression Region: Met1~His155. Tag Info: N-terminal His and GST Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0097