Learn More
Biomatik Corporation Recombinant Mouse Forkhead Box P3 (FOXP3)
Supplier:  Biomatik Corporation RPU41525
Encompass_Preferred
Recombinant Mouse Forkhead Box P3 (FOXP3) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Forkhead Box P3 (FOXP3). Target Synonyms: FOX-P3; AIID; DIETER; IPEX; JM2; PIDX; XPID; SCURFIN; Forkhead Box Protein P3; Immune Dysregulation,Polyendocrinopathy,Enteropathy,X-Linked. Accession Number: Q99JB6. Gene ID: 20371. Expression Region: Tyr190~Glu412. Tag Info: N-terminal His and GST Tag. Theoretical MW: 57kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9950