Learn More
Biomatik Corporation Recombinant Human Ferrochelatase (FECH)
Supplier:  Biomatik Corporation RPU51802
Encompass_Preferred
Recombinant Human Ferrochelatase (FECH) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Ferrochelatase (FECH). Target Synonyms: EPP; FCE; Protoheme Ferrolyase; Protoporphyria; Heme synthase; Protoheme ferro-lyase. Accession Number: P22830. Gene ID: 2235. Expression Region: Gly55~Leu423. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0163