Learn More
Biomatik Corporation Recombinant Human Phospholipase C Gamma 2, Phosphatidylinositol Specific (PLCg2)
Supplier:  Biomatik Corporation RPU52472
Encompass_Preferred
Recombinant Human Phospholipase C Gamma 2, Phosphatidylinositol Specific (PLCg2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Phospholipase C Gamma 2, Phosphatidylinositol Specific (PLCg2). Target Synonyms: PLC-IV; Phosphoinositide phospholipase C-gamma-2; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2. Accession Number: P16885. Gene ID: 5336. Expression Region: Leu930~Thr1152. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0192