Learn More
Biomatik Corporation Recombinant Human Voltage Dependent Anion Channel Protein 1 (VDAC1)
Supplier:  Biomatik Corporation RPU52906
Encompass_Preferred
Recombinant Human Voltage Dependent Anion Channel Protein 1 (VDAC1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Voltage Dependent Anion Channel Protein 1 (VDAC1). Target Synonyms: PORIN-31-HL; Porin 31HM; Plasmalemmal porin; Outer mitochondrial membrane protein porin 1; Voltage-dependent anion-selective channel protein 1. Accession Number: P21796. Expression Region: Ala2~Ala283. Tag Info: N-terminal His and GST Tag. Theoretical MW: 61kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0208