Learn More
Biomatik Corporation Recombinant Human Nuclear Mitotic Apparatus Protein 1 (NUMA1)
Supplier:  Biomatik Corporation RPU53413
Encompass_Preferred
Recombinant Human Nuclear Mitotic Apparatus Protein 1 (NUMA1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Nuclear Mitotic Apparatus Protein 1 (NUMA1). Target Synonyms: NMP22; NUMA; Nuclear matrix protein-22; SP-H antigen. Accession Number: Q14980. Gene ID: 4926. Expression Region: Phe1700~His2115. Tag Info: N-terminal His Tag. Theoretical MW: 50/56kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0223