Learn More
Biomatik Corporation Recombinant Human Alpha-Fetoprotein (AFP)
Supplier:  Biomatik Corporation RPU40203
Encompass_Preferred
Recombinant Human Alpha-Fetoprotein (AFP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Alpha-Fetoprotein (AFP). Target Synonyms: aFP; A-FP; FETA; HPAFP; Alpha-Fetoglobulin; Alpha-1-fetoprotein. Accession Number: P02771. Gene ID: 174. Expression Region: Ile31~Pro171. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9873