Learn More
Biomatik Corporation Recombinant Human Glutamate Cysteine Ligase, Catalytic (GCLC)
Supplier:  Biomatik Corporation RPU41638
Encompass_Preferred
Recombinant Human Glutamate Cysteine Ligase, Catalytic (GCLC) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glutamate Cysteine Ligase, Catalytic (GCLC). Target Synonyms: GCS; GLCL; GLCLC; Gamma-glutamylcysteine synthetase; Gamma-ECS. Accession Number: P48506. Gene ID: 2729. Expression Region: Met429~Asn637. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9957