Learn More
Biomatik Corporation Recombinant Mouse Asporin (ASPN)
Supplier:  Biomatik Corporation RPU53585
Encompass_Preferred
Recombinant Mouse Asporin (ASPN) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Asporin (ASPN). Target Synonyms: PLAP1; SLRR1C; Periodontal ligament-associated protein 1. Accession Number: Q99MQ4. Gene ID: 66695. Expression Region: Lys16~Lys373. Tag Info: N-terminal His and GST Tag. Theoretical MW: 71kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0229