Learn More
Biomatik Corporation Recombinant Human Cytoskeleton Associated Protein 4 (CKAP4)
Supplier:  Biomatik Corporation RPU56083
Encompass_Preferred
Recombinant Human Cytoskeleton Associated Protein 4 (CKAP4) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cytoskeleton Associated Protein 4 (CKAP4). Target Synonyms: P63; CLIMP-63; ERGIC-63; 63-kDa cytoskeleton-linking membrane protein. Accession Number: Q07065. Gene ID: 10970. Expression Region: His128~Val602. Tag Info: N-terminal His Tag. Theoretical MW: 58kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0326