Learn More
Biomatik Corporation Recombinant Rat Cholinergic Receptor, Nicotinic, Alpha 1 (CHRNa1)
Supplier:  Biomatik Corporation RPU56865
Encompass_Preferred
Recombinant Rat Cholinergic Receptor, Nicotinic, Alpha 1 (CHRNa1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Cholinergic Receptor, Nicotinic, Alpha 1 (CHRNa1). Target Synonyms: CHR-NA1; N-AChRA1; NAChRA1; ACHRA; ACHRD; CMS2A; FCCMS; SCCMS; Acetylcholine receptor subunit alpha; N-AChR-A1; Neuronal Acetylcholine Receptor Alpha 1,Muscle. Accession Number: P25108. Gene ID: 79557. Expression Region: Ser21~Phe245. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0342