Learn More
Biomatik Corporation Recombinant Mycobacterium tuberculosis ESAT-6-like protein EsxH (esxH), partial
Supplier:  Biomatik Corporation RPC20136
Encompass_Preferred
Recombinant Mycobacterium tuberculosis ESAT-6-like protein EsxH (esxH), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh). Target Name: esxH. Target Synonyms: 10 kDa antigen CFP7; CFP-7Low molecular weight protein antigen 7Protein TB10.4. Accession Number: P9WNK2. Expression Region: 2~96aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 26.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0379