Learn More
Biomatik Corporation Recombinant Human Actin Alpha 1, Cardiac Muscle (ACTC1)
Supplier:  Biomatik Corporation RPU40059
Encompass_Preferred
Recombinant Human Actin Alpha 1, Cardiac Muscle (ACTC1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Actin Alpha 1, Cardiac Muscle (Anti-ACTC1). Target Synonyms: ACTC; CMD1R; CMD1-R; ACT-a1. Accession Number: P68032. Gene ID: 70. Expression Region: Asp156~Gly368. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9865