Learn More
Biomatik Corporation Recombinant Human Sarcoplasmic/endoplasmic reticulum calcium ATPase 2 (ATP2A2), partial
Supplier:  Biomatik Corporation RPC20952
Encompass_Preferred
Recombinant Human Sarcoplasmic/endoplasmic reticulum calcium ATPase 2 (ATP2A2), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ATP2A2. Target Synonyms: Calcium pump 2; Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletal muscle isoform; Endoplasmic reticulum class 1/2 Ca(2+) ATPase. Accession Number: P16615. Expression Region: 314~756aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 52.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0435