Learn More
Biomatik Corporation Recombinant Mouse Cold-inducible RNA-binding protein (Cirbp)
Supplier:  Biomatik Corporation RPC21100
Encompass_Preferred
Recombinant Mouse Cold-inducible RNA-binding protein (Cirbp) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Cirbp. Target Synonyms: A18 hnRNPGlycine-rich RNA-binding protein CIRPCirp. Accession Number: P60824. Expression Region: 1~172aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 22.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0451