Learn More
Biomatik Corporation Recombinant Human NADPH:adrenodoxin oxidoreductase, mitochondrial (FDXR)
Supplier:  Biomatik Corporation RPC21311
Encompass_Preferred
Recombinant Human NADPH:adrenodoxin oxidoreductase, mitochondrial (FDXR) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: FDXR. Target Synonyms: Ferredoxin--NADP(+) reductase; Ferredoxin reductase. Accession Number: P22570. Expression Region: 33~451aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 61.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0476