Learn More
Biomatik Corporation Recombinant Human Heat Shock 70kDa Protein 5 (HSPA5)
Supplier:  Biomatik Corporation RPU50492
Encompass_Preferred
Recombinant Human Heat Shock 70kDa Protein 5 (HSPA5) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Heat Shock 70kDa Protein 5 (HSPA5). Target Synonyms: MIF2; BIP; GRP78; Immunoglobulin heavy chain-binding protein; Glucose Regulated Protein 78; Binding Immunoglobulin Protein; Endoplasmic reticulum lumenal Ca binding grp78. Accession Number: P11021. Gene ID: 3309. Expression Region: Gln261~Glu470. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0104