Learn More
Biomatik Corporation Recombinant Human Adenylate Cyclase 2, Brain (ADCY2)
Supplier:  Biomatik Corporation RPU40099
Encompass_Preferred
Recombinant Human Adenylate Cyclase 2, Brain (ADCY2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Adenylate Cyclase 2, Brain (ADCY2). Target Synonyms: HBAC2; ATP pyrophosphate-lyase 2; Adenylate cyclase type II. Accession Number: Q08462. Gene ID: 108. Expression Region: Ala294~Thr525. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9867