Learn More
Biomatik Corporation Recombinant Human Protein Tyrosine Phosphatase, Non Receptor Type 22 (PTPN22)
Supplier:  Biomatik Corporation RPU51004
Encompass_Preferred
Recombinant Human Protein Tyrosine Phosphatase, Non Receptor Type 22 (PTPN22) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Protein Tyrosine Phosphatase, Non Receptor Type 22 (PTPN22). Target Synonyms: LYP; PEP; Lyp1; Lyp2; PTPN8; Lymphoid phosphatase; Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP; PEST-domain phosphatase. Accession Number: Q9Y2R2. Gene ID: 26191. Expression Region: Met1~Arg233. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0119