Learn More
Biomatik Corporation Recombinant Human Brain Specific Angiogenesis Inhibitor 3 (BAI3)
Supplier:  Biomatik Corporation RPU51607
Encompass_Preferred
Recombinant Human Brain Specific Angiogenesis Inhibitor 3 (BAI3) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Brain Specific Angiogenesis Inhibitor 3 (BAI3). Accession Number: O60242. Gene ID: 577. Expression Region: Cys30~Pro508. Tag Info: N-terminal His Tag. Theoretical MW: 58kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0152