Learn More
Biomatik Corporation Recombinant Human N-Acylsphingosine Amidohydrolase 1 (ASAH1)
Supplier:  Biomatik Corporation RPU51792
Encompass_Preferred
Recombinant Human N-Acylsphingosine Amidohydrolase 1 (ASAH1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: N-Acylsphingosine Amidohydrolase 1 (ASAH1). Target Synonyms: AC; ASAH; PHP; PHP32; Acid Ceramidase; Acylsphingosine deacylase; Putative 32 kDa heart protein. Accession Number: Q13510. Gene ID: 427. Expression Region: Thr144~Trp395. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0161