Learn More
Biomatik Corporation Recombinant Human Alkaline Phosphatase, Placental Like Protein 2 (ALPPL2)
Supplier:  Biomatik Corporation RPU40159
Encompass_Preferred
Recombinant Human Alkaline Phosphatase, Placental Like Protein 2 (ALPPL2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Alkaline Phosphatase, Placental Like Protein 2 (ALPPL2). Target Synonyms: ALPG; GCAP; ALP-1; Alkaline phosphatase Nagao isozyme; Germ cell alkaline phosphatase; Placental alkaline phosphatase-like. Accession Number: P10696. Gene ID: 251. Expression Region: Ile20~Arg333. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9869