Learn More
Biomatik Corporation Recombinant Human Peptidoglycan Recognition Protein 1 (PGLYRP1)
Supplier:  Biomatik Corporation RPU51858
Encompass_Preferred
Recombinant Human Peptidoglycan Recognition Protein 1 (PGLYRP1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Peptidoglycan Recognition Protein 1 (PGLYRP1). Target Synonyms: PGLYRP; PGRP; PGRP-S; PGRPS; TAG7; TNFSF3L; Tumor Necrosis Factor Ligand Superfamily Member 3 Like; Peptidoglycan recognition protein short. Accession Number: O75594. Gene ID: 8993. Expression Region: Gln22~Pro196. Tag Info: N-terminal His Tag. Theoretical MW: 23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0167