Learn More
Biomatik Corporation Recombinant Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2B (GRIN2B)
Supplier:  Biomatik Corporation RPU51867
Encompass_Preferred
Recombinant Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2B (GRIN2B) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2B (GRIN2B). Target Synonyms: NMDAR2B; NR2B; hNR3; Glutamate [NMDA] receptor subunit epsilon-2; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 3. Accession Number: Q13224. Gene ID: 2904. Expression Region: Ile35~Asp557. Tag Info: N-terminal His and GST Tag. Theoretical MW: 89kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0169