Learn More
Biomatik Corporation Recombinant Human Thrombospondin 2 (THBS2)
Supplier:  Biomatik Corporation RPU51961
Encompass_Preferred
Recombinant Human Thrombospondin 2 (THBS2) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Thrombospondin 2 (THBS2). Target Synonyms: TSP2. Accession Number: P35442. Gene ID: 7058. Expression Region: Asp968~Ile1172. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0174