Learn More
Biomatik Corporation Recombinant Human Xanthine Dehydrogenase (XDH)
Supplier:  Biomatik Corporation RPU52169
Encompass_Preferred
Recombinant Human Xanthine Dehydrogenase (XDH) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Xanthine Dehydrogenase (XDH). Target Synonyms: XO; XOR; XOD; Xanthine Oxidase; NAD+-Xanthine Dehydrogenase; Xanthine-NAD+ Oxidoreductase; Xanthine/NAD+ Oxidoreductase; Xanthine Oxidoreductase. Accession Number: P47989. Gene ID: 7498. Expression Region: Met1~Gly300. Tag Info: N-terminal His and GST Tag. Theoretical MW: 63kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0179