Learn More
Biomatik Corporation Recombinant Human Suppressors Of Cytokine Signaling 2 (SOCS2)
Supplier:  Biomatik Corporation RPU52312
Encompass_Preferred
Recombinant Human Suppressors Of Cytokine Signaling 2 (SOCS2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Suppressors Of Cytokine Signaling 2 (SOCS2). Target Synonyms: CIS2; Cish2; SOCS-2; SSI-2; SSI2; STATI2; Cytokine-inducible SH2 protein 2; STAT-induced STAT inhibitor 2. Accession Number: O14508. Gene ID: 8835. Expression Region: Met1~Val198. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0181